Orders LinkContactUsLinkComponent
Hoffmann pinch cocks, Rotilabo®
20 mm
Katalog # ROTH1587.1
Marketsource
Leverandør:   Roth Carl
Hoffmann pinch cocks, Rotilabo®
Katalog # ROTH1587.1
Leverandør:  Roth Carl
Marketsource
Hoffmann pinch cocks, Rotilabo®
20 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Rotilabo®-tube clamping screws made of chrome-plated metal for fine regulation.
  • Tube clamping screws according to Hoffmann
  • Hinged bottom bar can be swing open
Specifikationer
  • Båndbredde:
    20 mm