Hoffmann pinch cocks, Rotilabo®
20 mm
Katalog # ROTH1587.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Rotilabo®-tube clamping screws made of chrome-plated metal for fine regulation.
- Tube clamping screws according to Hoffmann
- Hinged bottom bar can be swing open
Specifikationer
- Båndbredde:20 mm