Xylitol
Glass bottle for solids, 1 kg
Katalog # ROTH9711.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
- CAS nummer:87-99-0
- Formel:C₅H₁₂O₅
- MDL nummer:MFCD00064292
- Molecular Weight:152.15 g/mol
- Merck Index:13,10140
- Kogende Pt:495 °C (1013 hPa)
- Smeltende Pt:94 °C
- Tæthed:1,52 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-788-0
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg