Ethylenglycol
10 L
Katalog # ROTH9516.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 1,2-Ethandiol, Glykol
- CAS nummer:107-21-1
- Formel:HOCH₂CH₂OH
- MDL nummer:MFCD00002885
- Molecular Weight:62.07 g/mol
- Merck Index:13,03832
- Kogende Pt:196…198 °C (1013 hPa)
- Smeltende Pt:–13 °C
- Tæthed:1,115 g/cm³ (20 °C)
- Flash Pt:116 °C
- Opbevaringstemperatur:Køleskab
- EINECS:203-473-3
- NÅ:01-2119456816-28
Specifikationer
Specifikationer
- Pakn:10 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



