Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stivelse (from rice)

Stivelse (from rice)

Glass bottle for solids, 1 kg
Katalog # ROTH9368.1
Marketsource
Leverandør:   Roth Carl
Stivelse (from rice)
Katalog # ROTH9368.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  9005-25-8
Stivelse (from rice)

Glass bottle for solids, 1 kg

CAS nummer:  9005-25-8
Price
227,76 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
  • CAS nummer:9005-25-8
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-679-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer