Saltsyre 30%
Glass bottle, 1 L
Katalog # ROTH9280.3
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Hydrochloric acid solution, Muriatic acid
- CAS nummer:7647-01-0
- Formel:HCl
- MDL nummer:MFCD00011324
- Molecular Weight:36.46 g/mol
- Kogende Pt:110 °C (1013 hPa)
- Smeltende Pt:–30 °C
- Tæthed:1,18 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-595-7
- FN:1789
- ADR:8,II
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



