Order Entry
Denmark
Orders LinkContactUsLinkComponent
Natriumchlorid

Natriumchlorid

50 kg
Katalog # ROTH9265.4
Marketsource
Leverandør:   Roth Carl
Natriumchlorid
Katalog # ROTH9265.4
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7647-14-5
Natriumchlorid

50 kg

CAS nummer:  7647-14-5
Price
Spørg for pris
Log ind for tilgængelighed
1 * 50 kg
Product Details & Documents
Synonymer:  Saline, NaCl, Sea salt, Halite
  • CAS nummer:7647-14-5
  • Formel:NaCl
  • MDL nummer:MFCD00003477
  • Molecular Weight:58.44 g/mol
  • Merck Index:14,08599
  • Kogende Pt:1413 °C (1013 hPa)
  • Smeltende Pt:801 °C
  • Tæthed:2,16 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-598-3
Specifikationer

Specifikationer

  • Pakn:
    50 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.