Natriumchlorid
Glass bottle for solids, 1 kg
Katalog # ROTH9265.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Saline, NaCl, Sea salt, Halite
- CAS nummer:7647-14-5
- Formel:NaCl
- MDL nummer:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Kogende Pt:1413 °C (1013 hPa)
- Smeltende Pt:801 °C
- Tæthed:2,16 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-598-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg