Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumsulfat

Ammoniumsulfat

25 kg
Catalog # ROTH9218.5
Marketsource
Supplier:   Roth Carl
Ammoniumsulfat
Catalog # ROTH9218.5
Supplier:  Roth Carl
Marketsource
CAS Number:  7783-20-2
Ammoniumsulfat

25 kg

CAS Number:  7783-20-2
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
  • CAS Number:7783-20-2
  • Formula:(NH₄)₂SO₄
  • MDL Number:MFCD00003391
  • Molecular Weight:132.14 g/mol
  • Merck Index:13,00559
  • Melting Pt:336…339 °C
  • Density:1,78 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-984-1
  • REACH:01-2119455044-46
Specifications

Specifications

  • Pakn:
    25 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.