Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumsulfat

Ammoniumsulfat

5 kg
Katalog # ROTH9218.2
Marketsource
Leverandør:   Roth Carl
Ammoniumsulfat
Katalog # ROTH9218.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7783-20-2
Ammoniumsulfat

5 kg

CAS nummer:  7783-20-2
Price
Spørg for pris
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
Synonymer:  Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
  • CAS nummer:7783-20-2
  • Formel:(NH₄)₂SO₄
  • MDL nummer:MFCD00003391
  • Molecular Weight:132.14 g/mol
  • Merck Index:13,00559
  • Smeltende Pt:336…339 °C
  • Tæthed:1,78 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-984-1
  • NÅ:01-2119455044-46
Specifikationer

Specifikationer

  • Pakn:
    5 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.