ACES
Glass bottle for solids, 10 g
Katalog # ROTH9138.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS nummer:7365-82-4
- Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL nummer:MFCD00008030
- Molecular Weight:182.2 g/mol
- Merck Index:13,00037
- Smeltende Pt:270…295 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:230-908-4
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



