Orders LinkContactUsLinkComponent
Kaliumhydrogensulfat
Kaliumhydrogensulfat
Glass bottle for solids, 1 kg
Catalog # ROTHX989.2
Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Kaliumhydrogensulfat
Catalog # ROTHX989.2
Supplier:  Roth Carl
Marketsource
CAS Number:  7646-93-7
Kaliumhydrogensulfat
Glass bottle for solids, 1 kg
CAS Number:  7646-93-7
Price
369,65 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassium bisulphate
  • CAS Number:7646-93-7
  • Formula:KHSO₄
  • MDL Number:MFCD00011404
  • Molecular Weight:136.17 g/mol
  • Merck Index:13,07695
  • Melting Pt:214 °C
  • Density:2,32 g/cm³ (20 °C)
  • EINECS:231-594-1
  • UN:2509
  • ADR:8,II
Specifications
Specifications
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg