Natriumpyruvat
Glass bottle for solids, 25 g
Catalog # ROTH8793.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS Number:113-24-6
- Formula:CH₃COCOONa
- MDL Number:MFCD00002586
- Molecular Weight:110.04 g/mol
- Melting Pt:220…230 °C
- Density:1,718 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:204-024-4
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



