Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumsulfat

Natriumsulfat

25 kg
Katalog # ROTH8631.5
Marketsource
Leverandør:   Roth Carl
Natriumsulfat
Katalog # ROTH8631.5
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7757-82-6
Natriumsulfat
Natriumsulfat

25 kg

CAS nummer:  7757-82-6
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt
  • CAS nummer:7757-82-6
  • Formel:Na₂SO₄
  • MDL nummer:MFCD00003504
  • Molecular Weight:142.04 g/mol
  • Merck Index:12,08829
  • Kogende Pt:1429 °C (1013 hPa)
  • Smeltende Pt:888 °C
  • Tæthed:2,7 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-820-9
Specifikationer

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.