Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumcarbonat

Natriumcarbonat

25 kg
Katalog # ROTH8563.5
Irriterende
Marketsource
Leverandør:   Roth Carl
Natriumcarbonat
Katalog # ROTH8563.5
Leverandør:  Roth Carl
Marketsource
CAS nummer:  497-19-8
Natriumcarbonat
Natriumcarbonat

25 kg

CAS nummer:  497-19-8
Price
Spørg for pris
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  Calcined soda, Carbonic acid disodium salt
  • CAS nummer:497-19-8
  • Formel:Na₂CO₃
  • MDL nummer:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • Merck Index:14,08596
  • Kogende Pt:1600 °C (1013 hPa)
  • Smeltende Pt:854 °C
  • Tæthed:2,53 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:207-838-8
  • NÅ:01-2119485498-19
Specifikationer

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.