Methylethylketon
Glass bottle, 1 L
Katalog # ROTH8403.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Ethylmethylketon, Butanon
- CAS nummer:78-93-3
- Formel:C₂H₅COCH₃
- MDL nummer:MFCD00011648
- Molecular Weight:72.11 g/mol
- Merck Index:13,06097
- Kogende Pt:79,6 °C (1013 hPa)
- Smeltende Pt:–86 °C
- Tæthed:0,8054 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-159-0
- FN:1193
- ADR:3,II
- NÅ:01-2119457290-43
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



