Kaliumpermanganat
Glass bottle for solids, 1 kg
Katalog # ROTH8004.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Permanganic acid potassium salt
- CAS nummer:7722-64-7
- Formel:KMnO₄
- MDL nummer:MFCD00011364
- Molecular Weight:158.03 g/mol
- Merck Index:12,07824
- Smeltende Pt:> 240 °C
- Tæthed:2,7 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-760-3
- FN:1490
- ADR:5.1,II
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



