Kaliumdisulfit
10 kg
Katalog # ROTH7995.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
- CAS nummer:16731-55-8
- Formel:K₂O₅S₂
- MDL nummer:MFCD00167605
- Molecular Weight:222.33 g/mol
- Merck Index:13,07729
- Smeltende Pt:190 °C
- Tæthed:1,2 g/cm³ (20 °C)
- EINECS:240-795-3
Specifikationer
Specifikationer
- Pakn:10 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



