Kaliumdisulfit
Glass bottle for solids, 1 kg
Katalog # ROTH7995.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
- CAS nummer:16731-55-8
- Formel:K₂O₅S₂
- MDL nummer:MFCD00167605
- Molecular Weight:222.33 g/mol
- Merck Index:13,07729
- Smeltende Pt:190 °C
- Tæthed:1,2 g/cm³ (20 °C)
- EINECS:240-795-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



