Jod
Glass bottle for solids, 250 g
Katalog # ROTH7935.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Merck Index:13,05036
- Kogende Pt:184,3 °C (1013 hPa)
- Smeltende Pt:114 °C
- Tæthed:4,9 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-442-4
- FN:3495
- ADR:8,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 g