Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Saltsyre 25%

Saltsyre 25%

Glass bottle, 1 L
Catalog # ROTHX897.1
Corrosive
Irritant
Marketsource
Supplier:   Roth Carl
Saltsyre 25%
Catalog # ROTHX897.1
Supplier:  Roth Carl
Marketsource
CAS Number:  7647-01-0
Saltsyre 25%

Glass bottle, 1 L

CAS Number:  7647-01-0
Price
145,62 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1,18 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.