Eddikesyre 96% ≥96%
10 L
Catalog # ROTHX895.3
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Storage Temperature:Stue temperatur
- EINECS:200-580-7
- UN:2789
- ADR:8,II
Specifications
Specifications
- Pakn:10 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



