Eddikesyre 96% ≥96%
Glass bottle, 2,5 L
Catalog # ROTHX895.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Storage Temperature:Stue temperatur
- EINECS:200-580-7
- UN:2789
- ADR:8,II
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:2,5 L



