Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumacetat

Natriumacetat

25 kg
Katalog # ROTHX891.5
Marketsource
Leverandør:   Roth Carl
Natriumacetat
Katalog # ROTHX891.5
Leverandør:  Roth Carl
Marketsource
CAS nummer:  127-09-3
Natriumacetat

25 kg

CAS nummer:  127-09-3
Price
4.697,14 kr
Log ind for tilgængelighed
1 * 25 kg
Product Details & Documents
Synonymer:  Acetic acid sodium salt
  • CAS nummer:127-09-3
  • Formel:H₃CCOONa
  • MDL nummer:MFCD00012459
  • Molecular Weight:82.03 g/mol
  • Merck Index:12,08711
  • Kogende Pt:881,4 °C (1013 hPa)
  • Smeltende Pt:324 °C
  • Tæthed:1,528 g/cm³ (20 °C)
  • Flash Pt:>250 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-823-8
  • NÅ:01-2119485123-42

Documents

Specifikationer

Specifikationer

  • Pakn:
    25 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.