Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumsulfat

Kaliumsulfat

Glass bottle for solids, 2,5 kg
Katalog # ROTHX889.3
Marketsource
Leverandør:   Roth Carl
Kaliumsulfat
Katalog # ROTHX889.3
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7778-80-5
Kaliumsulfat

Glass bottle for solids, 2,5 kg

CAS nummer:  7778-80-5
Price
429,39 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
  • CAS nummer:7778-80-5
  • Formel:K₂SO₄
  • MDL nummer:MFCD00011388
  • Molecular Weight:174.26 g/mol
  • Merck Index:13,07759
  • Kogende Pt:1698 °C (1013 hPa)
  • Smeltende Pt:1069 °C
  • Tæthed:2,66 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-915-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2,5 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.