Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Transfer membranes, nitrocellulose

Transfer membranes, nitrocellulose

Nitrocellulose membranes, roll, pore size 0,45 µm, 200×3000 mm
Katalog # ROTH0041.1
Marketsource
Leverandør:   Roth Carl
Transfer membranes, nitrocellulose
Katalog # ROTH0041.1
Leverandør:  Roth Carl
Marketsource
Transfer membranes, nitrocellulose

Nitrocellulose membranes, roll, pore size 0,45 µm, 200×3000 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Nitrocellulose membranes, roll, pore size 0,45 µm
  • Dimensioner:
    200×3000 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.