Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Transfer membranes, nitrocellulose

Transfer membranes, nitrocellulose

Nitrocellulose membranes, sheets, pore size 0,45 µm, 102×133 mm
Katalog # ROTH0030.1
Marketsource
Leverandør:   Roth Carl
Transfer membranes, nitrocellulose
Katalog # ROTH0030.1
Leverandør:  Roth Carl
Marketsource
Transfer membranes, nitrocellulose

Nitrocellulose membranes, sheets, pore size 0,45 µm, 102×133 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Nitrocellulose membranes, sheets, pore size 0,45 µm
  • Dimensioner:
    102×133 mm
  • Seq:
    03