Jod
Glass bottle for solids, 1 kg
ROTHX864.3
Marketsource
- :7553-56-2
- :I₂
- :MFCD00011355
- :253.81 g/mol
- :13,05036
- :184,3 °C (1013 hPa)
- :114 °C
- :4,9 g/cm³ (20 °C)
- :Stue temperatur
- :231-442-4
- :3495
- :8,III
- Pakning:Glass bottle for solids
- Pakn:1 kg