Orders LinkContactUsLinkComponent
Jod
Glass bottle for solids, 250 g
  ROTHX864.2
Marketsource
 :   Roth Carl
Jod
  ROTHX864.2
 :  Roth Carl
Marketsource
 :  7553-56-2
Jod
Glass bottle for solids, 250 g
 :  7553-56-2
1.187,35 kr
1 * 250 g
  •  :7553-56-2
  •  :I₂
  •  :MFCD00011355
  •  :253.81 g/mol
  •  :13,05036
  •  :184,3 °C (1013 hPa)
  •  :114 °C
  •  :4,9 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :231-442-4
  •  :3495
  •  :8,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g