Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Citronsyre

Citronsyre

Glass bottle for solids, 2,5 kg
Katalog # ROTHX863.3
Irriterende
Marketsource
Leverandør:   Roth Carl
Citronsyre
Katalog # ROTHX863.3
Leverandør:  Roth Carl
Marketsource
CAS nummer:  77-92-9
Citronsyre
Citronsyre

Glass bottle for solids, 2,5 kg

CAS nummer:  77-92-9
Price
746,02 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
Synonymer:  2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
  • CAS nummer:77-92-9
  • Formel:HOC(COOH)(CH₂COOH)₂
  • MDL nummer:MFCD00011669
  • Molecular Weight:192.13 g/mol
  • Merck Index:13,02350
  • Kogende Pt:310 °C (1013 hPa)
  • Smeltende Pt:149…151 °C
  • Tæthed:1,665 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-069-1
  • NÅ:01-2119457026-42-0000
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2,5 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.