Citronsyre
Glass bottle for solids, 500 g
ROTHX863.1
Marketsource
: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
- :77-92-9
- :HOC(COOH)(CH₂COOH)₂
- :MFCD00011669
- :192.13 g/mol
- :13,02350
- :310 °C (1013 hPa)
- :149…151 °C
- :1,665 g/cm³ (20 °C)
- :Stue temperatur
- :201-069-1
- :01-2119457026-42-0000
- Pakning:Glass bottle for solids
- Pakn:500 g