Injection needles
24, 25 mm, 0,55 mm
Katalog # ROTHX804.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Suitable for all syringes with a Luer lock cone. Stainless steel with polished ground section.
Specifikationer
- Udv.Ø:0,55 mm
- Måler:24
- Længde:25 mm
- Seq:04