Saltsyre 32%
Glass bottle, 2,5 L
Katalog # ROTHP074.4
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Hydrochloric acid solution, Muriatic acid
- CAS nummer:7647-01-0
- Formel:HCl
- MDL nummer:MFCD00011324
- Molecular Weight:36.46 g/mol
- Kogende Pt:110 °C (1013 hPa)
- Smeltende Pt:–30 °C
- Tæthed:1,18 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-595-7
- FN:1789
- ADR:8,II
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



