Butylacetat
Glass bottle, 1 L
ROTHP036.1
Marketsource
: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
- :123-86-4
- :CH₃COO(CH₂)₃CH₃
- :MFCD00009445
- :116.16 g/mol
- :13,01534
- :126 °C (1013 hPa)
- :–76 °C
- :0,875 g/cm³ (20 °C)
- :25 °C
- :Stue temperatur
- :204-658-1
- :1123
- :3,III
- Pakning:Glass bottle
- Pakn:1 L