Kaliumsulfat
Glass bottle for solids, 1 kg
Katalog # ROTHP022.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
- CAS nummer:7778-80-5
- Formel:K₂SO₄
- MDL nummer:MFCD00011388
- Molecular Weight:174.26 g/mol
- Merck Index:13,07759
- Kogende Pt:1698 °C (1013 hPa)
- Smeltende Pt:1069 °C
- Tæthed:2,66 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-915-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



