Kaliumhydrogensulfat
Glass bottle for solids, 500 g
Katalog # ROTHP020.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Potassium bisulphate
- CAS nummer:7646-93-7
- Formel:KHSO₄
- MDL nummer:MFCD00011404
- Molecular Weight:136.17 g/mol
- Merck Index:13,07695
- Smeltende Pt:214 °C
- Tæthed:2,32 g/cm³ (20 °C)
- EINECS:231-594-1
- FN:2509
- ADR:8,II
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



