Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Borsyre

Borsyre

Glass bottle for solids, 1.000 g
Katalog # ROTHP010.2
Sundhed
Marketsource
Leverandør:   Roth Carl
Borsyre
Katalog # ROTHP010.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  10043-35-3
Borsyre
Borsyre

Glass bottle for solids, 1.000 g

CAS nummer:  10043-35-3
Price
339,78 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents
Synonymer:  Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
  • CAS nummer:10043-35-3
  • Formel:B(OH)₃
  • MDL nummer:MFCD00011337
  • Molecular Weight:61.83 g/mol
  • Merck Index:13,01326
  • Kogende Pt:185 °C (1013 hPa)
  • Smeltende Pt:160 °C
  • Tæthed:1,44 g/cm³ (25 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-139-2
  • NÅ:01-2119486683-25
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.