Borsyre
Glass bottle for solids, 500 g
Katalog # ROTHP010.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS nummer:10043-35-3
- Formel:B(OH)₃
- MDL nummer:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck Index:13,01326
- Kogende Pt:185 °C (1013 hPa)
- Smeltende Pt:160 °C
- Tæthed:1,44 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-139-2
- NÅ:01-2119486683-25
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



