Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylethylketon

Methylethylketon

Glass bottle, 1 L
Katalog # ROTHT920.1
Brandfarlig
Irriterende
Marketsource
Leverandør:   Roth Carl
Methylethylketon
Katalog # ROTHT920.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  78-93-3
Methylethylketon
Methylethylketon

Glass bottle, 1 L

CAS nummer:  78-93-3
Price
372,63 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Ethylmethylketon, Butanon
  • CAS nummer:78-93-3
  • Formel:C₂H₅COCH₃
  • MDL nummer:MFCD00011648
  • Molecular Weight:72.11 g/mol
  • Merck Index:13,06097
  • Kogende Pt:79,6 °C (1013 hPa)
  • Smeltende Pt:–86 °C
  • Tæthed:0,8054 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-159-0
  • FN:1193
  • ADR:3,II
  • NÅ:01-2119457290-43
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.