Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Diisopropylether

Diisopropylether

Glass bottle, 1 L
Katalog # ROTHT919.1
Brandfarlig
Irriterende
Marketsource
Leverandør:   Roth Carl
Diisopropylether
Katalog # ROTHT919.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  108-20-3
Diisopropylether
Diisopropylether

Glass bottle, 1 L

CAS nummer:  108-20-3
Price
335,30 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Isopropyl ether
  • CAS nummer:108-20-3
  • Formel:(CH₃)₂CHOCH(CH₃)₂
  • MDL nummer:MFCD00008880
  • Molecular Weight:102.18 g/mol
  • Merck Index:13,05232
  • Kogende Pt:68,5 °C (1013 hPa)
  • Smeltende Pt:–86 °C
  • Tæthed:0,726 g/cm³ (20 °C)
  • Flash Pt:–28 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-560-6
  • FN:1159
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.