Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Thiamin hydrochlorid

Glass bottle for solids, 100 g
Katalog # ROTHT911.2
Marketsource
Leverandør:   Roth Carl
Thiamin hydrochlorid
Katalog # ROTHT911.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  67-03-8
Thiamin hydrochlorid

Glass bottle for solids, 100 g

CAS nummer:  67-03-8
Price
566,79 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
  • CAS nummer:67-03-8
  • Formel:C₁₂H₁₈Cl₂N₄OS
  • MDL nummer:MFCD00012780
  • Molecular Weight:337.27 g/mol
  • Merck Index:13,09366
  • Smeltende Pt:252 °C
  • Tæthed:1,401 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-641-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer