Zinkchlorid
Glass bottle for solids, 1 kg
ROTHT887.2
Marketsource
: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
- :7646-85-7
- :ZnCl₂
- :MFCD00011295
- :136.3 g/mol
- :13,10185
- :732 °C (1013 hPa)
- :283 °C
- :2,91 g/cm³ (20 °C)
- :Stue temperatur
- :231-592-0
- :2331
- :8,III
- Pakning:Glass bottle for solids
- Pakn:1 kg