Orders LinkContactUsLinkComponent
Zinkchlorid
Glass bottle for solids, 1 kg
  ROTHT887.2
Marketsource
 :   Roth Carl
Zinkchlorid
  ROTHT887.2
 :  Roth Carl
Marketsource
 :  7646-85-7
Zinkchlorid
Glass bottle for solids, 1 kg
 :  7646-85-7
784,10 kr
1 * 1 kg
 :  Zinc dichloride, Dichlorozinc, Zinc (II) chloride
  •  :7646-85-7
  •  :ZnCl₂
  •  :MFCD00011295
  •  :136.3 g/mol
  •  :13,10185
  •  :732 °C (1013 hPa)
  •  :283 °C
  •  :2,91 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :231-592-0
  •  :2331
  •  :8,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg