Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinkchlorid

Zinkchlorid

Glass bottle for solids, 500 g
Katalog # ROTHT887.1
Ætsende
Irriterende
Miljø
Marketsource
Leverandør:   Roth Carl
Zinkchlorid
Katalog # ROTHT887.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7646-85-7
Zinkchlorid

Glass bottle for solids, 500 g

CAS nummer:  7646-85-7
Price
469,71 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Zinc dichloride, Dichlorozinc, Zinc (II) chloride
  • CAS nummer:7646-85-7
  • Formel:ZnCl₂
  • MDL nummer:MFCD00011295
  • Molecular Weight:136.3 g/mol
  • Merck Index:13,10185
  • Kogende Pt:732 °C (1013 hPa)
  • Smeltende Pt:283 °C
  • Tæthed:2,91 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-592-0
  • FN:2331
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.