Kaliumacetat
Glass bottle for solids, 1 kg
Katalog # ROTHT874.2
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Acetic acid potassium salt
- CAS nummer:127-08-2
- Formel:CH₃COOK
- MDL nummer:MFCD00012458
- Molecular Weight:98.14 g/mol
- Merck Index:13,07687
- Smeltende Pt:304 °C
- Tæthed:1,57 g/cm³ (25 °C)
- Flash Pt:>250 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-822-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg