Glycin
Glass bottle for solids, 1 kg
Catalog # ROTHT873.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:83 °C
- Density:1,64 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-272-2
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:1 kg



