Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kieselgel

Kieselgel

5 kg
Catalog # ROTHT858.2
Marketsource
Supplier:   Roth Carl
Kieselgel
Catalog # ROTHT858.2
Supplier:  Roth Carl
Marketsource
CAS Number:  63231-67-4
Kieselgel

5 kg

CAS Number:  63231-67-4
Price
1.262,03 kr
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:63231-67-4
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Boiling Pt:2230 °C (1013 hPa)
  • Melting Pt:1710 °C
  • Storage Temperature:Stue temperatur
  • REACH:01-2119379499-16
Specifications

Specifications

  • Pakn:
    5 kg

Specifications