Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Kieselgel

Glass bottle for solids, 1 kg
Katalog # ROTHT858.1
Marketsource
Leverandør:   Roth Carl
Kieselgel
Katalog # ROTHT858.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  63231-67-4
Kieselgel

Glass bottle for solids, 1 kg

CAS nummer:  63231-67-4
Price
384,58 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
  • CAS nummer:63231-67-4
  • Formel:SiO₂
  • MDL nummer:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Kogende Pt:2230 °C (1013 hPa)
  • Smeltende Pt:1710 °C
  • Opbevaringstemperatur:Stue temperatur
  • NÅ:01-2119379499-16
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.