Acetonitril
Glass bottle, 2,5 L
Katalog # ROTHT195.2
Marketsource
Leverandør: Roth Carl
Acetonitril
Katalog # ROTHT195.2
Leverandør: Roth Carl
Marketsource
CAS nummer: 75-05-8
Gå til:
Product Details & Documents
Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS nummer:75-05-8
- Formel:H₃CCN
- MDL nummer:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Kogende Pt:81,6 °C (1013 hPa)
- Smeltende Pt:–45,7 °C
- Tæthed:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-835-2
- FN:1648
- ADR:3,II
- NÅ:01-2119471307-38
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:ROTHT195.2
- Miljøcertificering:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



