Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyre 96% ≥96%

Eddikesyre 96% ≥96%

Glass bottle, 1 L
Katalog # ROTHT179.1
Brandfarlig
Ætsende
Marketsource
Leverandør:   Roth Carl
Eddikesyre 96% ≥96%
Katalog # ROTHT179.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  64-19-7
Eddikesyre 96% ≥96%
Eddikesyre 96% ≥96%

Glass bottle, 1 L

CAS nummer:  64-19-7
Price
235,23 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS nummer:64-19-7
  • Formel:H₃CCOOH
  • MDL nummer:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-580-7
  • FN:2789
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.