Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumpermanganat

Kaliumpermanganat

Glass bottle for solids, 1 kg
Katalog # ROTHP752.2
Oxiderende
Irriterende
Miljø
Marketsource
Leverandør:   Roth Carl
Kaliumpermanganat
Katalog # ROTHP752.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  7722-64-7
Kaliumpermanganat

Glass bottle for solids, 1 kg

CAS nummer:  7722-64-7
Price
813,97 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Permanganic acid potassium salt
  • CAS nummer:7722-64-7
  • Formel:KMnO₄
  • MDL nummer:MFCD00011364
  • Molecular Weight:158.03 g/mol
  • Merck Index:12,07824
  • Smeltende Pt:> 240 °C
  • Tæthed:2,7 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-760-3
  • FN:1490
  • ADR:5.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.