Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumhydrogenphthalat

Kaliumhydrogenphthalat

Glass bottle for solids, 1 kg
Katalog # ROTHP750.2
Marketsource
Leverandør:   Roth Carl
Kaliumhydrogenphthalat
Katalog # ROTHP750.2
Leverandør:  Roth Carl
Marketsource
CAS nummer:  877-24-7
Kaliumhydrogenphthalat
Kaliumhydrogenphthalat

Glass bottle for solids, 1 kg

CAS nummer:  877-24-7
Price
1.605,54 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
  • CAS nummer:877-24-7
  • Formel:HOOCC₆H₄COOK
  • MDL nummer:MFCD00013070
  • Molecular Weight:204.22 g/mol
  • Merck Index:13,07694
  • Smeltende Pt:295…300 °C
  • Tæthed:1,6362 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:212-889-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.